Learn More
Sino Biological Ferret TNF ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502372790
Encompass_Preferred
Each vial contains 200 ng of recombinant Ferret TNF-alpha / TNFA ELISA Pair Set. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 5000 pg/mL is recommended.
Catalog No. 50-237-2790 Supplier Sino Biological Supplier No. SEK600025
Each vial contains 200 ng of recombinant Ferret TNF-alpha / TNFA ELISA Pair Set. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 5000 pg/mL is recommended.