Learn More
Sino Biological Recombinant Anti-TNF-alpha Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 60002R30320
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Ferret TNF-alpha / TNFA / TNFSF2 ( Catalog#60002-FNAE; A3FBF1; Val77-Leu233).
Catalog No. 50-241-0002