Learn More
Sino Biological Anti-TNF-alpha Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 60002T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Ferret TNF-alpha / TNFA (Catalog#60002-FNAE; A3FBF1; Val77-Leu233). TNF-alpha / TNFA specific IgG was purified by Ferret TNF-alpha / TNFA affinity chromatography.
Catalog No. 50-240-8115