Learn More
Bioworld Ferric Ammonium Citrate, 100 g
Supplier:  Bioworld 406000011
Encompass_Preferred
Afood additive used as an acidity regulator.Used for water purification and printing.Used as a reducing agent of metal salts of low activity like gold and silverUsed to make cyanotype prints.Used in Kligler iron deeps to determine hydrogen sulfide production in microbial metabolism.Used as a contrast medium.Used as a hematinic.Synonym: Ammonium Iron(III) CitrateAppearance: Reddish-brown powderHS Code : 291815000IUPAC_Name: 2-Hydroxypropane-1,2,3-tricarboxylate, ammonium iron(3+) saltRTECS Number: GE7540000
Catalog No. 50-255-975