Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Ammonium Citrate Brown Gran Powder USP, DPL

Catalog No. 502985628
Encompass
Catalog No. 50-298-5628 Supplier Thermo Scientific Production Chemicals and Services Supplier No. D503007001
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

Molecular Weight (g/mol) 261.98 g/mol

Specifications

Boiling Point Undetermined
Quantity 25 kg
Percent Purity 16.5 to 18.5%
Grade USP
Physical Form Granular Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.