Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Ammonium Oxalate, Granular, Spectrum™ Chemical
SDP

Catalog No. 18620751 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-751 125 g
18-620-752 500 g
18-620-753 2.5 kg
18-620-754 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-620-751 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F1002125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Synonym: Ammonium Iron (III) Oxalate, Ammonium Ferrioxalate, Ammonium Ferric Oxalate

Ferric Ammonium Oxalate is a water soluble product used in the manufacturing of blueprint paper and for the chemical absorption on anodized aluminum, producing a range of gold colors from light champagne hues to dark gold finishes. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 13268-42-3
Molecular Formula C6H18FeN3O15
Molecular Weight (g/mol) 428.06
MDL Number MFCD00150137
InChI Key ARJSTCBPHIPXIJ-UHFFFAOYSA-K
IUPAC Name triammonium hexaoxo-1,4,6,9,10,13-hexaoxa-5-ferraspiro[4.4⁵.4⁵]tridecane-5,5,5-tris(ylium) trihydrate
SMILES [NH4+].[NH4+].[NH4+].O.O.O.O=C1O[Fe+3]234(OC1=O)OC(=O)C(=O)O2.O=C(O3)C(=O)O4

Specifications

Quantity 125 g
Packaging Amber Bottle
Chemical Name or Material Ferric Ammonium Oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.