Learn More
Electron Microscopy Sciences Ferric Ammonium Sulfate 2.5% Aqueous 500 ML
SDP
Catalog No. 5031883
Encompass_Preferred
Used in Woelcker's Method for Myelin Sheath for myelin sheath, glial cells and nucleoli of neurons. References: AFIP: Manual of Histological Staining Methods, 3rd ed., Ed. L. Luna: New York: McGraw Hill Publications, c. 1968, p. 206.
Catalog No. 50-318-83 Supplier Electron Microscopy Sciences Supplier No. 2668401
Add to Cart
Add to Cart
Used in Woelcker's Method for Myelin Sheath for myelin sheath, glial cells and nucleoli of neurons. References: AFIP: Manual of Histological Staining Methods, 3rd ed., Ed. L. Luna: New York: McGraw Hill Publications, c. 1968, p. 206.