Learn More
Electron Microscopy Sciences Ferric Ammonium Sulfate 5% Aqueous 500 ML
SDP
Catalog No. 5031795
Encompass_Preferred
Reference: For the staining of fibrin and platelets. Carstairs (1965).
Catalog No. 50-317-95 Supplier Electron Microscopy Sciences Supplier No. 2638101
Add to Cart
Add to Cart
Reference: For the staining of fibrin and platelets. Carstairs (1965).