Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Ammonium Sulfate Dodecahydrate, ACS Reagent, Reagents

Catalog No. 01337982
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-982 100 g
01-337-981 2.5 kg
01-337-980 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-982 Supplier Reagents Holdings Llc Supplier No. C1203500100C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferric Ammonium Sulfate Dodecahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.18
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym Iron(III) Ammonium Sulfate Dodecahydrate
IUPAC Name iron(3+) ammonium dodecahydrate disulfate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Boiling Point 230°C
Color Pale Violet
Physical Form Solid
Quantity 100 g
Percent Purity 98.5%
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.71
Chemical Name or Material Ferric Ammonium Sulfate Dodecahydrate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.