Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Ammonium Sulfate Solution, Reagents

Catalog No. 01335310
Change view
Click to view available options
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-335-310 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-310 Supplier Reagents Holdings Llc Supplier No. CF0002001C
 Restricted Item
Add to Cart
Add to Cart

Ferric Ammonium Sulfate Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Ferric Ammonium Sulfate Solution
Boiling Point 101°C
Color Amber
CAS 7732-18-5, 7697-37-2
Physical Form Liquid
Quantity 1 L
Synonym Iron(III) Ammonium Sulfate
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.3179
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.