Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Ammonium Sulfate TS (8% w/v), Reagents

Catalog No. 01335268
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-268 120 mL
01-335-267 1 L
01-335-266 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-335-268 Supplier Reagents Holdings Llc Supplier No. CF126100120A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferric Ammonium Sulfate TS (8% w/v), Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7783-83-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Iron(III) Ammonium Sulfate
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Orange
pH 2.5
Physical Form Liquid
Quantity 120 mL
Grade USP
Packaging Poly Bottle
Specific Gravity 1.0333
Chemical Name or Material Ferric Ammonium Sulfate TS (8% w/v)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.