Learn More
Medchemexpress LLC Ferric carboxymaltose | 53858-86-9 | 100 MG
SDP
Catalog No. 5000345585
Encompass_Preferred
Ferric carboxymaltose is an iron supplement designed for research use only. It is not intended for sale to patients or for medical applications. This product is provided as a solid, brown to reddish-brown in appearance, and should be handled by qualified scientists in equipped facilities.
  • For research use only
  • Not classified as a hazardous substance or mixture
  • Stable under recommended storage conditions
  • Recommended storage at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-45585 Supplier Medchemexpress LLC Supplier No. HY10876510MG
Ferric carboxymaltose is an iron supplement designed for research use only. It is not intended for sale to patients or for medical applications. This product is provided as a solid, brown to reddish-brown in appearance, and should be handled by qualified scientists in equipped facilities.
  • For research use only
  • Not classified as a hazardous substance or mixture
  • Stable under recommended storage conditions
  • Recommended storage at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month