Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Chloride, 0.2% w/v, Ricca Chemical
SDP

Catalog No. R31020001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R31020001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R31020001A Supplier Ricca Chemical Company Supplier No. R31020001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0 , 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer
PubChem CID 24380
ChEBI CHEBI:30808
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 100°C
Melting Point 0°C
Color Amber
CAS Min % 0.43, 99.37, 0.2
pH slightly acidic
CAS Max % 0.43, 99.37, 0.2
Physical Form Liquid
Quantity 1 L
Solubility Information Miscible in water
Percent Purity 0.2% W/V
Grade Laboratory
Packaging Natural Poly Bottle
Density 1.005
Chemical Name or Material Ferric Chloride, 0.2% w/v
Show More Show Less
ShelfLife 24 months
Recommended Storage Ambient
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.