Learn More
Electron Microscopy Sciences Ferric Chloride, 10% 500 ML
SDP
Catalog No. 5030089
Encompass_Preferred
Used in Modified Movat's Stain for connective tissue stains – Elastic tissue stains. See Movat, H.Z.;: Demonstration of all connective tissue elements in a single section. Arch Path., 60:289 – 295, 1955 or Roman N., et al: Orcein-hematoxylin in iodized ferric chloride as a stain for elastic tissue. Stain Technology, 42:199 –202, 1967
Catalog No. 50-300-89 Supplier Electron Microscopy Sciences Supplier No. 2638505
Used in Modified Movat's Stain for connective tissue stains – Elastic tissue stains. See Movat, H.Z.;: Demonstration of all connective tissue elements in a single section. Arch Path., 60:289 – 295, 1955 or Roman N., et al: Orcein-hematoxylin in iodized ferric chloride as a stain for elastic tissue. Stain Technology, 42:199 –202, 1967