Learn More
Electron Microscopy Sciences Ferric Chloride 2%, 500 ML
SDP
Catalog No. 5031787
Encompass_Preferred
Used in Verhoeff's Van-Gieson's (VVG Method) for staining elastic fibers, nuclei, collagen & other tissue elements. Reference is Sheehan, Dezna C., Hrapchack, Barbara B.: Theory and Practice of Histotechnology, 2nd ed. The C.V. Mosby C., St Louis, p. 196-197, c. 1980
Catalog No. 50-317-87 Supplier Electron Microscopy Sciences Supplier No. 2637404
Add to Cart
Add to Cart
Used in Verhoeff's Van-Gieson's (VVG Method) for staining elastic fibers, nuclei, collagen & other tissue elements. Reference is Sheehan, Dezna C., Hrapchack, Barbara B.: Theory and Practice of Histotechnology, 2nd ed. The C.V. Mosby C., St Louis, p. 196-197, c. 1980