Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Chloride, 40% (w/v), Ricca Chemical
SDP

Catalog No. 312716 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
312716 500 mL Natural Poly Bottle
3127-32 1 L Natural Poly Bottle
3127-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 312716 missing translation for 'mfr' Ricca Chemical Company missing translation for 'supplierNo' 312716
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10025-77-1 , 7732-18-5 , 7647-01-0
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer
PubChem CID 24380
ChEBI CHEBI:30808
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Color Amber
CAS Min % 27.78, 69.0, 0.82
CAS Max % 28.91, 72.0, 0.85
Percent Purity 40%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH Acidic
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 40% (w/v)
Chemical Name or Material Ferric Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.