Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Chloride, Anhydrous, MP Biomedicals

Catalog No. ICN223559 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN223559 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN223559 Supplier MP Biomedicals, Inc Supplier No. 0522355925
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anhydrous powder suitable for laboratory and research applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7705-08-0
Molecular Formula Cl3Fe
Molecular Weight (g/mol) 162.20
MDL Number MFCD00011005
InChI Key RBTARNINKXHZNM-UHFFFAOYSA-K
IUPAC Name iron(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Physical Form Powder
Quantity 25 g
Formula Weight 162.21
Percent Purity 98.8%
Chemical Name or Material Ferric Chloride, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.