Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Chloride Hexahydrate, Purified (Iron (III) Chloride Hexahydrate), Reagents

Catalog No. 01337251
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-251 100 g
01-337-249 2 kg
01-337-248 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-251 Supplier Reagents Holdings Llc Supplier No. C2204000100C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferric Chloride Hexahydrate, Purified (Iron (III) Chloride Hexahydrate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Iron(III) Chloride Hexahydrate
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 280°C
Melting Point 37°C
Color Brownish-Yellow
Physical Form Solid
Quantity 100 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 1.82
Chemical Name or Material Ferric Chloride Hexahydrate, Purified (Iron (III) Chloride Hexahydrate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.