Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Chloride, MP Biomedicals

Catalog No. MP522355980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP522355980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP522355980 Supplier MP Biomedicals, Inc Supplier No. 0522355980
Add to Cart
Add to Cart

  • Ferric Chloride is used in photoengraving, photography, the manufacture of pigments and ink, the chlorination of silver and copper ores, and as a mordant in dyeing and printing textiles
  • It is also utilized in the synthesis of organic compounds
  • FeCl3 is used in the Mayer's tannic acid/ferric chloride method for staining tissue and their observation by light microscopy
  • Studies of iron uptake and of apoptosis in cultured cells have utilized FeCl3 as an iron source
  • The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resitance properties

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7705-08-0
Molecular Formula Cl3Fe
Molecular Weight (g/mol) 162.20
MDL Number MFCD00011005
InChI Key RBTARNINKXHZNM-UHFFFAOYSA-K
Synonym Chlorure Perrique, Iron (III) Chloride, Iron Trichloride, Perchlorure de Fer, Molysite, Ferric chloride
IUPAC Name iron(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 315°C (literature)
Melting Point 306°C (literature)
Color Black
Physical Form Powder
Quantity 100 g
Linear Formula FeCl3
Solubility Information Soluble in water.
Formula Weight 162.2
Chemical Name or Material FERRIC CHLORIDE
Recommended Storage Room Temperature, desiccate, store under nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.