Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Chloride Solution, Reagents

Catalog No. 01335309
Change view
Click to view available options
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-335-309 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-309 Supplier Reagents Holdings Llc Supplier No. CF0004001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferric Chloride Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Ferric Chloride Solution
Boiling Point 102°C
Color Orange
CAS 7732-18-5, 10025-77-1
Physical Form Liquid
Quantity 1 L
Molecular Formula FeCl3/H2O
Synonym Iron(III) Chloride
Percent Purity 20%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.099
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.