Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Chloride TS 9% w/v, Reagents

Numéro de catalogue. 01335253
Modifier l'affichage
Click to view available options
Quantity:
120 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
01-335-253 120 mL
01-335-252 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 01-335-253 Fournisseur Reagents Holdings Llc Code fournisseur CF128800120C
Ajouter au panier
Ajouter au panier

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Ferric Chloride TS 9% w/v, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Identifiants chimiques

CAS 7732-18-5 , 10025-77-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Iron(III) Chloride
IUPAC Name water
SMILES O

Spécifications

Boiling Point 100°C
Color Orange/Brown
pH <7
Physical Form Liquid
Quantity 120 mL
Grade USP
Packaging Glass Bottle
Specific Gravity 1.1
Chemical Name or Material Ferric Chloride TS 9% w/v
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.