Learn More
Medchemexpress LLC Ferric citrate | 3522-50-7 | 244.94 | 100 MG
SDP
Catalog No. 5000416119
Encompass_Preferred
Ferric citrate (Iron(III) citrate) is an orally active iron supplement and an efficacious phosphate binder. It is used for research related to iron deficiency anemia and chronic kidney disease (CKD).
  • Orally active iron supplement.
  • Efficacious phosphate binder.
  • Can be used for iron deficiency anemia and chronic kidney disease (CKD) research.
  • Reduces fibroblast growth factor 23 levels and improves renal and cardiac function in a mouse model of chronic kidney disease.
  • Increases iron stores and reduces intravenous iron and erythropoietin-stimulating agent use while maintaining hemoglobin.
  • Can increase transferrin saturation, serum ferritin, and hemoglobin.
Catalog No. 50-004-16119 Supplier Medchemexpress LLC Supplier No. HYN1428C100MG
Ferric citrate (Iron(III) citrate) is an orally active iron supplement and an efficacious phosphate binder. It is used for research related to iron deficiency anemia and chronic kidney disease (CKD).
  • Orally active iron supplement.
  • Efficacious phosphate binder.
  • Can be used for iron deficiency anemia and chronic kidney disease (CKD) research.
  • Reduces fibroblast growth factor 23 levels and improves renal and cardiac function in a mouse model of chronic kidney disease.
  • Increases iron stores and reduces intravenous iron and erythropoietin-stimulating agent use while maintaining hemoglobin.
  • Can increase transferrin saturation, serum ferritin, and hemoglobin.