Learn More
Sigma Aldrich Fine Chemicals Biosciences Ferric citrate, 3522-50-7, MFCD00013098, 250 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences F3388250G
Encompass
Empirical Formula (Hill Notation): C6H5FeO7, Molecular Weight: 244.94, BioReagent, suitable for cell culture, Synonym: Iron(III) citrate hydrate, Iron(III) citrate.
Catalog No. NC0554657