Learn More
Medchemexpress LLC Ferric citrate hydrate | 2338-05-8 | MFCD00013098 | 99.8% | C6H8FeO7 | 25 G
SDP
Catalog No. 5000358948
Encompass_Preferred
Ferric citrate hydrate is an orally active iron supplement that also functions as an efficacious phosphate binder. It is intended for research use only.
  • Orally active iron supplement
  • Efficacious phosphate binder
  • Used for research related to iron deficiency anemia
  • Used for research related to chronic kidney disease (CKD)
Catalog No. 50-003-58948 Supplier Medchemexpress LLC Supplier No. 25GHY47030
Ferric citrate hydrate is an orally active iron supplement that also functions as an efficacious phosphate binder. It is intended for research use only.
  • Orally active iron supplement
  • Efficacious phosphate binder
  • Used for research related to iron deficiency anemia
  • Used for research related to chronic kidney disease (CKD)