Learn More
Alkali Scientific Ferric Citrate Hydrate [Fe (III) Citrate Hydrate], 50 Grams
SDP
Catalog No. 5000660499
Encompass_Preferred
Ferric Citrate Hydrate (Fe III citrate hydrate) is a key iron source, supplied in a 50g bottle for laboratory use. It is commonly used as a supplement in cell culture media, particularly in the study of iron metabolism and the effects of iron on cellular processes. Ferric Citrate is essential in research involving iron-related enzymes, heme production, and the regulation of cellular oxidative stress. In pharmaceutical and biological research, it is used to simulate iron-deficient conditions or to provide the necessary iron for cell growth. It also plays a crucial role in microbiological studies related to iron uptake and bacterial growth in different environments.
Catalog No. 50-006-60499 Supplier Alkali Scientific Supplier No. FER001CP
Ferric Citrate Hydrate (Fe III citrate hydrate) is a key iron source, supplied in a 50g bottle for laboratory use. It is commonly used as a supplement in cell culture media, particularly in the study of iron metabolism and the effects of iron on cellular processes. Ferric Citrate is essential in research involving iron-related enzymes, heme production, and the regulation of cellular oxidative stress. In pharmaceutical and biological research, it is used to simulate iron-deficient conditions or to provide the necessary iron for cell growth. It also plays a crucial role in microbiological studies related to iron uptake and bacterial growth in different environments.