Learn More
Hach Company Ferric Ion Solution, 100 mL
Supplier:  Hach Company 2212242
Encompass
For colorimetric chloride determination by the mercuric thiocyanate method. Hach method 8113. Range: 0.1 to 25.0 mg/L Cl- 100 mL bottle
Catalog No. 50-199-5561