Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ferric maltol | 33725-54-1 | MFCD33402141 | 99.9% | 431.15 g·mol⁻¹ | C18H15FeO9 | 25 MG
SDP

Supplier:  Medchemexpress LLC HY10801725MG

Encompass_Preferred

Ferric maltol is an orally active iron(III) complex used as an oral iron supplement and a research reagent to study iron deficiency anemia, including cases associated with inflammatory bowel disease. It is supplied for laboratory use with reported high purities and is available in both solid and solution formats.

  • High reported purity: 99.86%-99.93%.
  • Molecular formula C18H15FeO9.
  • Molecular weight 431.15 g·mol⁻¹.
  • Supplied in solid form and as DMSO solutions.
  • Available in multiple pack sizes, including 25 mg.
  • Commonly used to study oral iron delivery and iron deficiency anemia.

Catalog No. 50-002-42157


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.