Learn More
Ferric Nitrate Nonahydrate, Purified (Iron (III) Nitrate Nonahydrate), Reagents
Specifications
Specifications
| Boiling Point | 125°C |
| Melting Point | 47.2°C |
| Color | Pale Mauve |
| Physical Form | Solid |
| Quantity | 500 g |
| Percent Purity | 98% |
| Grade | Pure |
| Packaging | Glass Bottle |
| Specific Gravity | 1.68 |
| Chemical Name or Material | Ferric Nitrate Nonahydrate, Purified (Iron (III) Nitrate Nonahydrate) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.