Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Nitrate Solution, Stock, 202 g/L in dilute Nitric Acid, Ricca Chemical
SDP

Catalog No. R313400020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
20 L
Packaging:
Amber Poly Bottle
Cubitainer
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R313400020F 20 L Cubitainer
3134-16 500 mL Amber Poly Bottle
3134-32 1 L Amber Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R313400020F Supplier Ricca Chemical Company Supplier No. 31345
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7697-37-2 , 7782-61-8 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Brown
Grade Laboratory
Packaging Cubitainer
pH <1
Quantity 20 L
Physical Form Liquid
Chemical Name or Material Ferric Nitrate Solution, Stock Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.