Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ferric nitrilotriacetate | 16448-54-7 | 98.0% | 243.96 | C6H6FeNO6 | 5 MG
SDP

Catalog No. 5000221117
Encompass_Preferred

Ferric nitrilotriacetate is an iron-nitrilotriacetate complex used as a biochemical reagent in research, commonly employed in studies of metal-mediated oxidative processes and assay development. Supplied as a light yellow solid with high purity, it is intended for laboratory research use.

  • High purity (98.0%) suitable for research applications.
  • Light yellow solid, easy to handle and weigh.
  • Formula C6H6FeNO6 and molecular weight 243.96 for accurate calculations.
  • Available in small quantities for assay development and testing.
  • Soluble in DMSO and available as a 10 mM solution for convenience.

Catalog No. 50-002-21117 Supplier Medchemexpress LLC Supplier No. HY1451575MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferric nitrilotriacetate is an iron-nitrilotriacetate complex used as a biochemical reagent in research, commonly employed in studies of metal-mediated oxidative processes and assay development. Supplied as a light yellow solid with high purity, it is intended for laboratory research use.

  • High purity (98.0%) suitable for research applications.
  • Light yellow solid, easy to handle and weigh.
  • Formula C6H6FeNO6 and molecular weight 243.96 for accurate calculations.
  • Available in small quantities for assay development and testing.
  • Soluble in DMSO and available as a 10 mM solution for convenience.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.