Learn More
Medchemexpress LLC Ferric nitrilotriacetate | 16448-54-7 | 98.0% | C6H6FeNO6 | 1 ML
SDP
Catalog No. 5000355423
Encompass_Preferred
Ferric nitrilotriacetate (Fe-NTA), a complexation of nitriloacetic acid with iron, is a highly reactive compound used to induce degenerative disorders through oxidative stress (OS). It is also employed in various studies to induce hyperglycemia, glycosuria, and both renal and liver carcinogenesis.
  • Induces degenerative disorders through oxidative stress.
  • Employed in studies to induce hyperglycemia, glycosuria, and both renal and liver carcinogenesis.
  • Can be used in animal modeling to create renal tumor and diabetes models.
  • Induces oxidative stress leading to lipid peroxidation of renal cell membranes and DNA damage.
  • Causes significant iron deposition in the pancreas, leading to oxidative stress and damage to pancreatic islet β-cells.
  • For research use only.
Catalog No. 50-003-55423 Supplier Medchemexpress LLC Supplier No. HY14515710MM/1ML
Ferric nitrilotriacetate (Fe-NTA), a complexation of nitriloacetic acid with iron, is a highly reactive compound used to induce degenerative disorders through oxidative stress (OS). It is also employed in various studies to induce hyperglycemia, glycosuria, and both renal and liver carcinogenesis.
  • Induces degenerative disorders through oxidative stress.
  • Employed in studies to induce hyperglycemia, glycosuria, and both renal and liver carcinogenesis.
  • Can be used in animal modeling to create renal tumor and diabetes models.
  • Induces oxidative stress leading to lipid peroxidation of renal cell membranes and DNA damage.
  • Causes significant iron deposition in the pancreas, leading to oxidative stress and damage to pancreatic islet β-cells.
  • For research use only.