Learn More
Medchemexpress LLC Ferric nitrilotriacetate | 16448-54-7 | 98.0% | C6H6FeNO6 | 100 MG
SDP
Catalog No. 5000355752
Encompass_Preferred
Ferric nitrilotriacetate (Fe-NTA) is a complexation of nitriloacetic acid with iron. It is a highly reactive compound used to induce degenerative disorders through oxidative stress (OS). This product is also utilized in several studies to induce hyperglycemia, glycosuria, and both renal and liver carcinogenesis, and is intended for research use only.
  • Induces degenerative disorders via oxidative stress
  • Used to induce hyperglycemia, glycosuria, and renal and liver carcinogenesis
  • Can be used for animal modeling to create renal tumor and diabetes models
  • Induces lipid peroxidation of renal cell membranes and DNA damage
  • Causes significant iron deposition in the pancreas leading to oxidative stress and damage to pancreatic islet beta-cells
Catalog No. 50-003-55752 Supplier Medchemexpress LLC Supplier No. HY145157100MG
Ferric nitrilotriacetate (Fe-NTA) is a complexation of nitriloacetic acid with iron. It is a highly reactive compound used to induce degenerative disorders through oxidative stress (OS). This product is also utilized in several studies to induce hyperglycemia, glycosuria, and both renal and liver carcinogenesis, and is intended for research use only.
  • Induces degenerative disorders via oxidative stress
  • Used to induce hyperglycemia, glycosuria, and renal and liver carcinogenesis
  • Can be used for animal modeling to create renal tumor and diabetes models
  • Induces lipid peroxidation of renal cell membranes and DNA damage
  • Causes significant iron deposition in the pancreas leading to oxidative stress and damage to pancreatic islet beta-cells