Learn More
Medchemexpress LLC Ferric nitrilotriacetate | 16448-54-7 | 98.0% | C6H6FeNO6 | 10MG
SDP
Catalog No. 5000261903
Encompass_Preferred
Ferric nitrilotriacetate (Fe-NTA) is the iron complex of nitrilotriacetic acid used as a reactive oxidant in biological and toxicology research. It induces oxidative stress and is employed to model renal and hepatic carcinogenesis and to provoke metabolic changes such as hyperglycemia and glycosuria in experimental systems. The compound is supplied as a light yellow solid with high specified purity and recommended inert storage conditions.
  • Used to induce oxidative stress in cellular and animal models.
  • Suitable for studies of renal and liver carcinogenesis.
  • Allows experimental induction of hyperglycemia and glycosuria.
  • High purity for reproducible experimental results.
  • Light yellow solid, convenient for handling and weighing.
  • Stable when stored under inert atmosphere at recommended temperatures.
Catalog No. 50-002-61903 Supplier Medchemexpress LLC Supplier No. HY14515710MG
Ferric nitrilotriacetate (Fe-NTA) is the iron complex of nitrilotriacetic acid used as a reactive oxidant in biological and toxicology research. It induces oxidative stress and is employed to model renal and hepatic carcinogenesis and to provoke metabolic changes such as hyperglycemia and glycosuria in experimental systems. The compound is supplied as a light yellow solid with high specified purity and recommended inert storage conditions.
  • Used to induce oxidative stress in cellular and animal models.
  • Suitable for studies of renal and liver carcinogenesis.
  • Allows experimental induction of hyperglycemia and glycosuria.
  • High purity for reproducible experimental results.
  • Light yellow solid, convenient for handling and weighing.
  • Stable when stored under inert atmosphere at recommended temperatures.