Learn More
Medchemexpress LLC Ferric nitrilotriacetate | 16448-54-7 | 98.0% | C6H6FeNO6 | 25 MG
SDP
Catalog No. 5000353532
Encompass_Preferred
Ferric nitrilotriacetate (Fe-NTA) is a complexation of nitriloacetic acid with iron. This highly reactive compound is utilized in research to induce degenerative disorders through oxidative stress, and is also employed in various studies to induce hyperglycemia, glycosuria, and both renal and liver carcinogenesis.
  • Induces oxidative stress, useful for studying degenerative disorders.
  • Serves as a tool to induce hyperglycemia and glycosuria.
  • Facilitates research into renal and liver carcinogenesis.
  • Can be used in animal modeling to create renal tumor and diabetes models.
  • Provides a standardized method for researchers to study disease mechanisms related to iron overload and oxidative damage.
Catalog No. 50-003-53532 Supplier Medchemexpress LLC Supplier No. 25MGHY145157
Ferric nitrilotriacetate (Fe-NTA) is a complexation of nitriloacetic acid with iron. This highly reactive compound is utilized in research to induce degenerative disorders through oxidative stress, and is also employed in various studies to induce hyperglycemia, glycosuria, and both renal and liver carcinogenesis.
  • Induces oxidative stress, useful for studying degenerative disorders.
  • Serves as a tool to induce hyperglycemia and glycosuria.
  • Facilitates research into renal and liver carcinogenesis.
  • Can be used in animal modeling to create renal tumor and diabetes models.
  • Provides a standardized method for researchers to study disease mechanisms related to iron overload and oxidative damage.