Learn More
Medchemexpress LLC Ferric nitrilotriacetate | 16448-54-7 | 98.0% | C6H6FeNO6 | 50 MG
SDP
Catalog No. 5000355419
Encompass_Preferred
Ferric nitrilotriacetate (Fe-NTA) is a complexation of nitriloacetic acid with iron. It is a highly reactive compound employed to induce degenerative disorders through oxidative stress (OS). Additionally, Ferric nitrilotriacetate is utilized in various studies to induce hyperglycemia, glycosuria, and both renal and liver carcinogenesis. It is intended for research use only and not sold to patients.
  • Used to induce degenerative disorders through oxidative stress (OS)
  • Utilized in various studies to induce hyperglycemia, glycosuria, and both renal and liver carcinogenesis
  • Can be used in animal modeling to create renal tumor and diabetes models
  • Deposits heavily in the liver and pancreas after intraperitoneal injection into mice
Catalog No. 50-003-55419 Supplier Medchemexpress LLC Supplier No. HY14515750MG
Ferric nitrilotriacetate (Fe-NTA) is a complexation of nitriloacetic acid with iron. It is a highly reactive compound employed to induce degenerative disorders through oxidative stress (OS). Additionally, Ferric nitrilotriacetate is utilized in various studies to induce hyperglycemia, glycosuria, and both renal and liver carcinogenesis. It is intended for research use only and not sold to patients.
  • Used to induce degenerative disorders through oxidative stress (OS)
  • Utilized in various studies to induce hyperglycemia, glycosuria, and both renal and liver carcinogenesis
  • Can be used in animal modeling to create renal tumor and diabetes models
  • Deposits heavily in the liver and pancreas after intraperitoneal injection into mice