Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Oxide, Red, Technical (Iron (III) Oxide), Reagents

Catalog No. 01337244
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-244 500 g
01-337-246 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-244 Supplier Reagents Holdings Llc Supplier No. C2205000500A5
Only null left
Add to Cart
Add to Cart

Ferric Oxide, Red, Technical (Iron (III) Oxide), Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
Synonym Iron(III) Oxide, Rust
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O

Specifications

Boiling Point 3414°C
Melting Point 1565°C
Color Orange/Red
Physical Form Solid
Quantity 500 g
Grade Technical
Packaging Poly Bottle
Specific Gravity 5.25
Chemical Name or Material Ferric Oxide, Red, Technical (Iron (III) Oxide)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.