Learn More
Medchemexpress LLC Ferric oxide, yellow | 51274-00-1 | MFCD00011008 | Fe2O3·H2O | 50 G
SDP
Catalog No. 5000376895
Encompass_Preferred
Ferric oxide, yellow is an excipient that can be used as a colorant in pharmaceutical preparations. It is generally an inactive ingredient used to improve stability, solubility, and processability, and can influence the absorption, distribution, metabolism, and elimination processes of co-administered drugs.
  • Provides color uniformity
  • Masks odors
  • Functions as a light blocker to protect photosensitive components
  • Can serve as an iron supplement in some nutritional supplements
Catalog No. 50-003-76895 Supplier Medchemexpress LLC Supplier No. HY15469650G
Ferric oxide, yellow is an excipient that can be used as a colorant in pharmaceutical preparations. It is generally an inactive ingredient used to improve stability, solubility, and processability, and can influence the absorption, distribution, metabolism, and elimination processes of co-administered drugs.
  • Provides color uniformity
  • Masks odors
  • Functions as a light blocker to protect photosensitive components
  • Can serve as an iron supplement in some nutritional supplements