Learn More
Medchemexpress LLC Ferric pyrophosphate | 10058-44-3 | 25 G
SDP
Catalog No. 5000414428
Encompass_Preferred
Ferric pyrophosphate is a biological material or organic compound commonly used in life science research. It appears as an off-white to light yellow solid.
  • Appears as an off-white to light yellow solid
  • Powder storage at -20°C for 3 years
  • In solvent storage at -80°C for 6 months, or -20°C for 1 month
Catalog No. 50-004-14428 Supplier Medchemexpress LLC Supplier No. 25GHYB1648
Ferric pyrophosphate is a biological material or organic compound commonly used in life science research. It appears as an off-white to light yellow solid.
  • Appears as an off-white to light yellow solid
  • Powder storage at -20°C for 3 years
  • In solvent storage at -80°C for 6 months, or -20°C for 1 month