Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Sulfate AR Powder, Macron Fine Chemicals™

Catalog No. MK5036500 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass
STAKMOR
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-5036-500 500 g Glass
MK-5036-212 2.5 kg STAKMOR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MK-5036-500 Supplier Avantor Supplier No. 503612
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15244-10-7
Molecular Formula Fe2H2O13S3
Molecular Weight (g/mol) 417.87
InChI Key VXWSFRMTBJZULV-UHFFFAOYSA-H
PubChem CID 167265
IUPAC Name diiron(3+) hydrate trisulfate
SMILES O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Physical Form Powder
Quantity 500 g
Packaging Glass
Chemical Name or Material Ferric Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.