Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ferric Sulfate, Hydrated
SCI_ED

Catalog No. p-4416383 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25322A 500 g Reagent Grade
S25322 100 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25322A
Only null left
Add to Cart
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10028-22-5
Molecular Formula Fe2O12S3
Molecular Weight (g/mol) 399.858
InChI Key RUTXIHLAWFEWGM-UHFFFAOYSA-H
Synonym ferric sulfate, iron iii sulfate, iron persulfate, iron tersulfate, diiron tris sulphate, diiron trisulfate, ferric persulfate, ferric tersulfate, iron sesquisulfate, ferric sesquisulfate
PubChem CID 24826
ChEBI CHEBI:53438
IUPAC Name iron(3+);trisulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3]

Specifications

Color Gray-Yellow
Quantity 500 g
Linear Formula FeSO4 · H2O
Formula Weight 399.87
Grade Reagent
Packaging Reagent Grade
Density 3.097g/mL
Chemical Name or Material Ferric Sulfate, Hydrated
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.