Learn More
Key Scientific Products DROPIT - PYZ REAGENT (FES04), Ferric Sulfate Reagent, 5 per Pack
SDP
Catalog No. NC9020308
Encompass_Preferred
Ferric sulfate reagent is pure ferric sulfate in tablet form with no added ingredients. The reagent is used with K1537 PYZ or comparable tests requiring ferric sulfate reagent. This reagent normally has a very short shelf life but these stable tablets allow fresh reagent as needed. Tablets are sold in packs of 5 dropper vials, each of which are to be reconstituted with water making only 3 ml of fresh usable reagent.
Catalog No. NC9020308 Supplier Key Scientific Products Supplier No. K980195
Ferric sulfate reagent is pure ferric sulfate in tablet form with no added ingredients. The reagent is used with K1537 PYZ or comparable tests requiring ferric sulfate reagent. This reagent normally has a very short shelf life but these stable tablets allow fresh reagent as needed. Tablets are sold in packs of 5 dropper vials, each of which are to be reconstituted with water making only 3 ml of fresh usable reagent.