Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Sulfate, Technical, Spectrum™ Chemical
SDP

Catalog No. 18602686 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-686 100 g Amber Glass Bottle
18-602-683 500 g Amber Glass Bottle
18-602-684 2.5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-602-686 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F1047100GM

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

F1047, 15244-10-7, Fe2(SO4)3·xH2O

Spectrum™ Chemical Ferric Sulfate, Technical is used to prepare ferric subsulfate solutions. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 15244-10-7
Molecular Formula Fe2H2O13S3
Molecular Weight (g/mol) 417.87
InChI Key VXWSFRMTBJZULV-UHFFFAOYSA-H
IUPAC Name diiron(3+) hydrate trisulfate
SMILES O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 100 g
Grade Technical
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.