Learn More
Cayman Chemical FerrIoxamIn E 10mg
SDP
Catalog No. 502939166
Encompass_Preferred
A bacterial siderophore; has been used as a growth enriching agent in the detection of Salmonella in food and industrial applications
Catalog No. 50-293-9166 Supplier Cayman Chemical Supplier No. 2727510 MG
A bacterial siderophore; has been used as a growth enriching agent in the detection of Salmonella in food and industrial applications