Learn More
Crystal Chem Inc Ferritin ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80964
Encompass
The Ferritin ELISA is a highly sensitive assay that is used to quantify levels of human ferritin in serum. Highlights: Kit uses sample size of only 20 µL. High sensitivity of 7.5 ng/mL. Wide dynamic range of 10 - 800 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7869