Learn More
Sigma Aldrich Fine Chemicals Biosciences ferritin from human liver
Catalog No. 502902661
Encompass
ferritin comprises 24-subunit and belongs to the ferritin-like diiron-carboxylate protein superfamily. ferritin exists as heavy (H) and light (L) subunits. Liver ferritin is majorly L subunit and the spleen has the H submit. The L subunit is mapped to human chromosome 9q13.33 and the H subunit 11q12.3.
Catalog No. 50-290-2661 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F67541VL
ferritin comprises 24-subunit and belongs to the ferritin-like diiron-carboxylate protein superfamily. ferritin exists as heavy (H) and light (L) subunits. Liver ferritin is majorly L subunit and the spleen has the H submit. The L subunit is mapped to human chromosome 9q13.33 and the H subunit 11q12.3.