Learn More
Apexbio Technology LLC ferritin heavy chain fragment [Multiple species] 5mg
SDP
Catalog No. 5000096454
Encompass_Preferred
Ferritin heavy chain fragment is a protein subunit fragment associated with iron storage and metabolism It is derived from H-ferritin a component predominantly expressed in heart kidney placenta and various tumor cells and forms part of the larger 24-subunit ferritin complex The ferritin heavy chain exhibits ferroxidase activity catalyzing the oxidation of Fe to Fe thereby sequestering intracellular iron and reducing the pool of catalytically active free iron Based on its biochemical attributes ferritin heavy chain fragment is widely utilized as a research tool or biomarker in studies investigating iron metabolism disorders iron overload diseases tumor biology inflammation and immune modulation in oncology
Catalog No. 50-000-96454 Supplier Apexbio Technology LLC Supplier No. A10695
Ferritin heavy chain fragment is a protein subunit fragment associated with iron storage and metabolism It is derived from H-ferritin a component predominantly expressed in heart kidney placenta and various tumor cells and forms part of the larger 24-subunit ferritin complex The ferritin heavy chain exhibits ferroxidase activity catalyzing the oxidation of Fe to Fe thereby sequestering intracellular iron and reducing the pool of catalytically active free iron Based on its biochemical attributes ferritin heavy chain fragment is widely utilized as a research tool or biomarker in studies investigating iron metabolism disorders iron overload diseases tumor biology inflammation and immune modulation in oncology