Learn More
ABclonal Technology Ferritin Heavy Chain Rabbit
SDP
Catalog No. 5000012975
Encompass_Preferred
This gene encodes the heavy subunit of ferritin the major intracellular iron storage protein in prokaryotes and eukaryotes It is composed of 24 subunits of the heavy and light ferritin chains Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues A major function of ferritin is the storage of iron in a soluble and nontoxic state Defects in ferritin proteins are associated with several neurodegenerative diseases This gene has multiple pseudogenes Several alternatively spliced transcript variants have been observed but their biological validity has not been determined
Catalog No. 50-000-12975 Supplier ABclonal Technology Supplier No. A25458PM20UL
This gene encodes the heavy subunit of ferritin the major intracellular iron storage protein in prokaryotes and eukaryotes It is composed of 24 subunits of the heavy and light ferritin chains Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues A major function of ferritin is the storage of iron in a soluble and nontoxic state Defects in ferritin proteins are associated with several neurodegenerative diseases This gene has multiple pseudogenes Several alternatively spliced transcript variants have been observed but their biological validity has not been determined