Learn More
Cayman Chemical FerrItIn Heavy ChaInhumn200ug
SDP
Catalog No. 502950783
Encompass_Preferred
Source: Recombinant human ferritin heavy chain expressed in E. coli • Amino acids: 1-183 (full length) • MW: 21.2 kDa
Catalog No. 50-295-0783 Supplier Cayman Chemical Supplier No. 32033200 UG
Source: Recombinant human ferritin heavy chain expressed in E. coli • Amino acids: 1-183 (full length) • MW: 21.2 kDa