Learn More
Cayman Chemical FerrItIn LIght ChaInhumn100ug
SDP
Catalog No. 502946166
Encompass_Preferred
Source: Recombinant human N-terminal His-tagged ferritin light chain expressed in E. coli • Amino acids: 1-175 • MW: 22.1 kDa
Catalog No. 50-294-6166 Supplier Cayman Chemical Supplier No. 32032100 UG
Source: Recombinant human N-terminal His-tagged ferritin light chain expressed in E. coli • Amino acids: 1-175 • MW: 22.1 kDa