Learn More
Electron Microscopy Sciences Ferritin on a Grid
SDP
Supplier:  Electron Microscopy Sciences 80042
Encompass_Preferred
Calibration specimens for TEM and STEM. Resolution of the quad structure of this molecule indicates an instrument resolution better than 1.25nm with a photomicrograph negative magnification of at least 100,000x.
Catalog No. 50-192-8652